Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D4A
Entrez Gene ID
63898
Gene Name
SH2 domain containing 4A
Gene Aliases
PPP1R38, SH2A
Chromosome Location
Chr.8: 19313443 - 19396218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174159.1 NP_001167630.1 3 - 4 646 101
NM_001174160.1 NP_001167631.1 2 - 3 411 101
NM_022071.3 NP_071354.2 3 - 4 796 101
XM_011544610.1 XP_011542912.1 3 - 4 923 101
XM_017013729.1 XP_016869218.1 3 - 4 645 101
GenBank mRNA AK024620.1 - 1 - 2 65 101
AK024799.1 - 3 - 4 674 101
AK225052.1 - 1 - 2 65 101
AK293301.1 - 2 - 3 287 101
BC082982.1 - 3 - 4 646 101

View Full Product Details