The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001174159.1 | NP_001167630.1 | 7 - 8 | 1231 | 88 |
NM_001174160.1 | NP_001167631.1 | 6 - 7 | 996 | 88 | |
NM_022071.3 | NP_071354.2 | 7 - 8 | 1381 | 88 | |
XM_011544610.1 | XP_011542912.1 | 6 - 7 | 1427 | 88 | |
XM_017013729.1 | XP_016869218.1 | 6 - 7 | 1149 | 88 | |
GenBank mRNA | AK024799.1 | - | 7 - 8 | 1259 | 88 |
AK293301.1 | - | 6 - 7 | 872 | 88 | |
AY190323.1 | - | 3 - 4 | 410 | 88 | |
BC014525.2 | - | 4 - 5 | 577 | 88 | |
BC067117.1 | - | 4 - 5 | 591 | 88 | |
BC082982.1 | - | 7 - 8 | 1231 | 88 |