Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SH2D4A
Entrez Gene ID
63898
Gene Name
SH2 domain containing 4A
Gene Aliases
PPP1R38, SH2A
Chromosome Location
Chr.8: 19313443 - 19396218 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174159.1 NP_001167630.1 8 - 9 1356 81
NM_001174160.1 NP_001167631.1 7 - 8 1121 81
NM_022071.3 NP_071354.2 8 - 9 1506 81
XM_011544610.1 XP_011542912.1 7 - 8 1552 81
XM_017013729.1 XP_016869218.1 7 - 8 1274 81
GenBank mRNA AK024620.1 - 6 - 7 775 81
AK024799.1 - 8 - 9 1384 81
AK225052.1 - 6 - 7 775 81
AK293301.1 - 7 - 8 997 81
AY190323.1 - 4 - 5 81
BC014525.2 - 5 - 6 702 81
BC067117.1 - 5 - 6 716 81
BC082982.1 - 8 - 9 1356 81

View Full Product Details