The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_031947.3 | NP_114153.1 | 1 - 1 | 1342 | 61 |
GenBank mRNA | AF332005.1 | - | 1 - 1 | 1144 | 61 |
AF378119.1 | - | 1 - 1 | 1324 | 61 | |
AY208943.1 | - | 1 - 1 | 1252 | 61 |