Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACRC
Entrez Gene ID
93953
Gene Name
acidic repeat containing
Gene Aliases
NAAR1
Chromosome Location
Chr.X: 71578024 - 71613583 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052957.4 NP_443189.1 6 - 7 726 76
XM_006724716.3 XP_006724779.1 6 - 7 279 76
XM_011531063.2 XP_011529365.1 5 - 6 546 76
XM_011531064.1 XP_011529366.1 6 - 7 329 76
XM_011531066.2 XP_011529368.1 3 - 4 465 76
XM_011531067.2 XP_011529369.1 3 - 4 465 76
XM_017029965.1 XP_016885454.1 7 - 8 380 76
GenBank mRNA AJ311392.1 - 5 - 6 227 76
AK097433.1 - 6 - 7 340 76
AK127607.1 - 5 - 6 762 76
BC136259.1 - 6 - 7 294 76

View Full Product Details