The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030785.3 | NP_110412.1 | 3 - 4 | 1824 | 64 |
XM_011527351.1 | XP_011525653.1 | 3 - 4 | 1800 | 64 | |
GenBank mRNA | AK093068.1 | - | 2 - 3 | 1094 | 64 |
AK098837.1 | - | 3 - 4 | 1769 | 64 | |
AK223347.1 | - | 3 - 4 | 1769 | 64 | |
AL136761.1 | - | 3 - 4 | 1796 | 64 | |
BC057785.1 | - | 3 - 4 | 1652 | 64 | |
BC136351.1 | - | 3 - 4 | 1705 | 64 | |
BC136352.1 | - | 3 - 4 | 1705 | 64 |