The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005242.2 | NP_001005242.2 | 9 - 10 | 2125 | 72 |
NM_004572.3 | NP_004563.2 | 10 - 11 | 2257 | 72 | |
GenBank mRNA | AK299139.1 | - | 5 - 6 | 800 | 72 |
BC094762.1 | - | 9 - 10 | 2048 | 72 | |
BC126199.1 | - | 9 - 10 | 2054 | 72 | |
BC143966.1 | - | 9 - 10 | 2054 | 72 | |
EU520483.1 | - | 10 - 11 | 2142 | 72 | |
EU520484.1 | - | 10 - 11 | 2142 | 72 | |
EU520485.1 | - | 10 - 11 | 2140 | 72 | |
EU520486.1 | - | 10 - 11 | 2142 | 72 | |
X97675.1 | - | 10 - 11 | 2167 | 72 |