Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PKP2
Entrez Gene ID
5318
Gene Name
plakophilin 2
Gene Aliases
ARVD9
Chromosome Location
Chr.12: 32790746 - 32896846 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001005242.2 NP_001005242.2 1 - 2 337 153
NM_004572.3 NP_004563.2 1 - 2 337 153
GenBank mRNA BC094762.1 - 1 - 2 260 153
BC126199.1 - 1 - 2 266 153
BC143966.1 - 1 - 2 266 153
BX956935.1 - 1 - 2 266 153
EU520483.1 - 1 - 2 222 153
EU520484.1 - 1 - 2 222 153
EU520485.1 - 1 - 2 220 151
EU520486.1 - 1 - 2 222 153
X97675.1 - 1 - 2 247 153

View Full Product Details