Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PPME1
Entrez Gene ID
51400
Gene Name
protein phosphatase methylesterase 1
Gene Aliases
PME-1
Chromosome Location
Chr.11: 74170891 - 74254703 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271593.1 NP_001258522.1 8 - 9 1071 109
NM_016147.2 NP_057231.1 8 - 9 1071 109
XM_017017913.1 XP_016873402.1 8 - 9 1243 109
GenBank mRNA AF111853.1 - 2 - 3 289 109
AF157028.1 - 8 - 9 811 109
AK001381.1 - 8 - 9 807 109
AK022725.1 - 8 - 9 881 109
AK025414.1 - 2 - 3 171 109
AK225647.1 - 8 - 9 812 109
BC003046.1 - 8 - 9 822 109
BC050705.2 - 8 - 9 818 109

View Full Product Details