The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038603.2 | NP_001033692.2 | 2 - 3 | 1214 | 86 |
XM_005248445.4 | XP_005248502.1 | 2 - 3 | 1508 | 86 | |
XM_005248446.4 | XP_005248503.1 | 2 - 3 | 1302 | 86 | |
GenBank mRNA | AB219936.1 | - | 1 - 2 | 1140 | 86 |
AB219937.1 | - | 1 - 2 | 1140 | 86 | |
AK055094.1 | - | 2 - 3 | 1210 | 86 | |
AK292962.1 | - | 2 - 3 | 1198 | 86 | |
BC033689.1 | - | 2 - 3 | 1199 | 86 | |
DQ682656.1 | - | 2 - 3 | 1199 | 86 | |
DQ682658.1 | - | 2 - 3 | 810 | 86 |