The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022569.2 | NP_072091.1 | 3 - 4 | 1748 | 87 |
XM_017008545.1 | XP_016864034.1 | 2 - 3 | 746 | 87 | |
GenBank mRNA | AB036429.1 | - | 3 - 4 | 1748 | 87 |
AK289670.1 | - | 3 - 4 | 1726 | 87 |