Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CCDC78
Entrez Gene ID
124093
Gene Name
coiled-coil domain containing 78
Gene Aliases
C16orf25, CNM4, JFP10, hsCCDC78
Chromosome Location
Chr.16: 722578 - 726880 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011522361.1 XP_011520663.1 8 - 9 1736 97
XM_017022929.1 XP_016878418.1 8 - 9 1736 97
XM_017022931.1 XP_016878420.1 1 - 2 989 97
XM_017022932.1 XP_016878421.1 4 - 5 830 97
GenBank mRNA AK057178.1 - 11 - 12 1544 97

View Full Product Details