The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098475.1 | NP_001091945.1 | 10 - 11 | 1638 | 115 |
NM_182499.3 | NP_872305.3 | 10 - 11 | 1638 | 115 | |
XM_006711156.2 | XP_006711219.1 | 5 - 6 | 1268 | 115 | |
XM_011509152.1 | XP_011507454.1 | 11 - 12 | 1027 | 115 | |
XM_011509153.1 | XP_011507455.1 | 11 - 12 | 1027 | 115 | |
XM_011509154.1 | XP_011507456.1 | 11 - 12 | 985 | 115 | |
XM_011509155.1 | XP_011507457.1 | 11 - 12 | 1027 | 115 | |
XM_011509156.1 | XP_011507458.1 | 10 - 11 | 952 | 115 | |
XM_011509157.1 | XP_011507459.1 | 10 - 11 | 952 | 115 | |
XM_011509158.2 | XP_011507460.1 | 6 - 7 | 742 | 115 | |
XM_011509159.2 | XP_011507461.1 | 6 - 7 | 733 | 115 | |
XM_011509160.2 | XP_011507462.1 | 6 - 7 | 778 | 115 | |
XM_011509161.1 | XP_011507463.1 | 6 - 7 | 746 | 115 | |
XM_017000255.1 | XP_016855744.1 | 5 - 6 | 672 | 115 | |
GenBank mRNA | AK302144.1 | - | 10 - 11 | 885 | 115 |
AL713777.1 | - | 5 - 6 | 543 | 115 | |
AY358195.1 | - | 4 - 5 | 407 | 115 | |
BC044864.1 | - | 10 - 11 | 1637 | 115 | |
BC100895.1 | - | 5 - 6 | 516 | 115 | |
BC100896.1 | - | 5 - 6 | 516 | 115 | |
BC100897.2 | - | 5 - 6 | 516 | 115 | |
BC100898.2 | - | 5 - 6 | 516 | 115 |