Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
Chromosome Location
Chr.11: 44066118 - 44084237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127219.1 NP_001120691.1 11 - 12 1110 60
NM_032592.3 NP_115981.1 11 - 12 1406 60
XM_005253170.3 XP_005253227.2 11 - 12 1549 60
XM_005253172.2 XP_005253229.1 8 - 9 637 60
XM_006718348.2 XP_006718411.1 11 - 12 858 60
XM_011520406.2 XP_011518708.1 11 - 12 1552 60
XM_011520407.1 XP_011518709.1 11 - 12 902 60
XM_011520410.2 XP_011518712.1 11 - 12 1803 60
GenBank mRNA AK057649.1 - 11 - 12 1400 60
AL833533.1 - 3 - 4 2403 60
AY026508.1 - 11 - 12 1361 60
BC020197.1 - 11 - 12 1380 60
KU178811.1 - 10 - 11 966 60
KU178812.1 - 10 - 11 966 60
KU178813.1 - 9 - 10 906 60

View Full Product Details