Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
Chromosome Location
Chr.11: 44066118 - 44084237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127219.1 NP_001120691.1 13 - 14 1388 115
NM_032592.3 NP_115981.1 13 - 14 1684 115
XM_005253170.3 XP_005253227.2 13 - 14 1827 115
XM_005253172.2 XP_005253229.1 10 - 11 915 115
XM_006718348.2 XP_006718411.1 13 - 14 1136 115
XM_011520407.1 XP_011518709.1 13 - 14 1180 115
XM_011520410.2 XP_011518712.1 13 - 14 2081 115
GenBank mRNA AK057649.1 - 13 - 14 1678 115
AY026508.1 - 13 - 14 1639 115
BC020197.1 - 13 - 14 1658 115
KU178811.1 - 12 - 13 1244 115
KU178812.1 - 12 - 13 1244 115
KU178813.1 - 11 - 12 1184 115

View Full Product Details