Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACCS
Entrez Gene ID
84680
Gene Name
1-aminocyclopropane-1-carboxylate synthase homolog (inactive)
Gene Aliases
ACS, PHACS
Chromosome Location
Chr.11: 44066118 - 44084237 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127219.1 NP_001120691.1 14 - 15 1551 86
NM_032592.3 NP_115981.1 14 - 15 1847 86
XM_005253170.3 XP_005253227.2 14 - 15 1990 86
XM_005253172.2 XP_005253229.1 11 - 12 1078 86
XM_006718348.2 XP_006718411.1 14 - 15 1299 86
XM_011520407.1 XP_011518709.1 14 - 15 1343 86
XM_011520410.2 XP_011518712.1 14 - 15 2244 86
GenBank mRNA AK057649.1 - 14 - 15 1841 86
AL833533.1 - 6 - 7 2844 86
AY026508.1 - 14 - 15 1802 86
BC020197.1 - 14 - 15 1821 86
KU178811.1 - 13 - 14 1407 86
KU178813.1 - 12 - 13 1347 86

View Full Product Details