Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RORA
Entrez Gene ID
6095
Gene Name
RAR related orphan receptor A
Gene Aliases
NR1F1, ROR1, ROR2, ROR3, RZR-ALPHA, RZRA
Chromosome Location
Chr.15: 60488284 - 61229303 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002943.3 NP_002934.1 2 - 3 430 112
GenBank mRNA U04899.1 - 2 - 3 352 112

View Full Product Details