The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012175.3 | NP_036307.2 | 5 - 6 | 704 | 78 |
NM_033406.2 | NP_208385.1 | 5 - 6 | 704 | 78 | |
XM_011519980.1 | XP_011518282.1 | 5 - 6 | 710 | 78 | |
XM_011519981.2 | XP_011518283.1 | 5 - 6 | 710 | 78 | |
XM_011519982.1 | XP_011518284.1 | 6 - 7 | 764 | 78 | |
XM_017017533.1 | XP_016873022.1 | 6 - 7 | 764 | 78 | |
GenBank mRNA | AB209039.1 | - | 4 - 5 | 4089 | 78 |
AF176702.1 | - | 5 - 6 | 661 | 78 | |
AK001943.1 | - | 5 - 6 | 695 | 78 | |
AK128454.1 | - | 5 - 6 | 682 | 78 | |
AL832826.1 | - | 4 - 5 | 1376 | 78 | |
BC039291.1 | - | 5 - 6 | 704 | 78 | |
BC046110.1 | - | 5 - 6 | 690 | 78 |