Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
KIAA0930
Entrez Gene ID
23313
Gene Name
KIAA0930
Gene Aliases
C22orf9, LSC3
Chromosome Location
Chr.22: 45192242 - 45240769 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001009880.1 NP_001009880.1 9 - 10 1242 57
NM_015264.1 NP_056079.1 9 - 10 1314 57
XM_011530017.2 XP_011528319.1 9 - 10 1277 57
XM_017028672.1 XP_016884161.1 9 - 10 1286 57
GenBank mRNA AB023147.1 - 8 - 9 1010 57
AK025608.1 - 7 - 8 831 57
AK094937.1 - 10 - 11 1325 57
AK124648.1 - 9 - 10 1322 57
AK297058.1 - 9 - 10 1294 57
AK309015.1 - 8 - 9 1366 57
AK315883.1 - 9 - 10 1226 57
AY007105.1 - 2 - 3 196 57
BC016882.1 - 4 - 5 572 57
BC038414.1 - 8 - 9 1389 57
BC053595.1 - 9 - 10 1236 57
CR456402.1 - 9 - 10 1179 57

View Full Product Details