The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003026.3 | NP_003017.1 | 6 - 7 | 907 | 115 |
XM_011518005.2 | XP_011516307.1 | 6 - 7 | 3085 | 115 | |
GenBank mRNA | AB206840.1 | - | 6 - 7 | 618 | 115 |
AF036268.1 | - | 6 - 7 | 665 | 115 | |
AK293592.1 | - | 6 - 7 | 709 | 115 | |
AK302273.1 | - | 7 - 8 | 760 | 115 | |
AK312400.1 | - | 6 - 7 | 738 | 115 | |
BC032825.2 | - | 6 - 7 | 794 | 115 | |
BT019682.1 | - | 6 - 7 | 618 | 115 | |
BX538074.1 | - | 3 - 4 | 609 | 115 | |
CR542086.1 | - | 6 - 7 | 618 | 115 | |
CR542102.1 | - | 6 - 7 | 618 | 115 | |
X99657.1 | - | 6 - 7 | 627 | 115 |