Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EZR
Entrez Gene ID
7430
Gene Name
ezrin
Gene Aliases
CVIL, CVL, HEL-S-105, VIL2
Chromosome Location
Chr.6: 158765741 - 158819424 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111077.1 NP_001104547.1 7 - 8 879 134
NM_003379.4 NP_003370.2 6 - 7 913 134
XM_011536110.1 XP_011534412.1 3 - 4 290 134
GenBank mRNA AF187552.1 - 1 - 2 126 134
AF188896.1 - 1 - 2 112 134
AK296738.1 - 7 - 8 708 134
AK299456.1 - 5 - 6 742 134
AK312597.1 - 7 - 8 823 134
AL162086.1 - 7 - 8 831 134
BC013903.2 - 6 - 7 830 134
BC068458.1 - 6 - 7 839 134
FJ224318.1 - 6 - 7 815 134
J05021.1 - 6 - 7 774 134
X51521.1 - 6 - 7 815 134

View Full Product Details