Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RSPH3
Entrez Gene ID
83861
Gene Name
radial spoke 3 homolog
Gene Aliases
CILD32, RSHL2, RSP3, dJ111C20.1
Chromosome Location
Chr.6: 158947012 - 159000432 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031924.4 NP_114130.3 6 - 7 1468 118
XM_005267153.4 XP_005267210.1 4 - 5 1446 118
XM_017011347.1 XP_016866836.1 5 - 6 789 118
GenBank mRNA AF353618.1 - 6 - 7 1067 118
AK057655.1 - 4 - 5 795 118
AK057931.1 - 6 - 7 1489 118
BC050604.1 - 6 - 7 1468 118

View Full Product Details