The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_005253263.2 | XP_005253320.1 | 1 - 2 | 584 | 80 |
XM_005253265.2 | XP_005253322.1 | 1 - 2 | 596 | 80 | |
XM_006718394.2 | XP_006718457.1 | 1 - 2 | 584 | 80 | |
XM_011520493.1 | XP_011518795.1 | 1 - 2 | 584 | 80 | |
XM_011520500.1 | XP_011518802.1 | 1 - 2 | 584 | 80 | |
XM_011520501.1 | XP_011518803.1 | 1 - 2 | 584 | 80 | |
XM_011520502.1 | XP_011518804.1 | 1 - 2 | 584 | 80 | |
XM_011520503.1 | XP_011518805.1 | 1 - 2 | 584 | 80 | |
XM_017018602.1 | XP_016874091.1 | 1 - 2 | 596 | 80 | |
XM_017018603.1 | XP_016874092.1 | 1 - 2 | 596 | 80 | |
XM_017018604.1 | XP_016874093.1 | 1 - 2 | 584 | 80 | |
XM_017018609.1 | XP_016874098.1 | 1 - 2 | 584 | 80 | |
XM_017018612.1 | XP_016874101.1 | 1 - 2 | 596 | 80 | |
XM_017018613.1 | XP_016874102.1 | 1 - 2 | 584 | 80 | |
XM_017018618.1 | XP_016874107.1 | 1 - 2 | 584 | 80 | |
XM_017018620.1 | XP_016874109.1 | 1 - 2 | 584 | 80 |