The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291278.1 | NP_001278207.1 | 5 - 6 | 576 | 79 |
NM_001291279.1 | NP_001278208.1 | 6 - 7 | 719 | 79 | |
NM_080475.3 | NP_536723.2 | 8 - 9 | 1019 | 79 | |
XM_011526251.1 | XP_011524553.1 | 7 - 8 | 1060 | 79 | |
GenBank mRNA | AF419953.1 | - | 6 - 7 | 774 | 79 |
AF419954.1 | - | 6 - 7 | 774 | 79 | |
AF419955.1 | - | 6 - 7 | 774 | 79 | |
AK292775.1 | - | 7 - 8 | 836 | 79 | |
AY739645.1 | - | 8 - 9 | 1018 | 79 | |
AY792324.1 | - | 4 - 5 | 513 | 79 | |
BC069596.1 | - | 6 - 7 | 774 | 79 | |
BC130370.1 | - | 6 - 7 | 800 | 79 | |
BC130372.1 | - | 6 - 7 | 800 | 79 |