The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286457.1 | NP_001273386.1 | 6 - 7 | 789 | 95 |
NM_001286458.1 | NP_001273387.1 | 7 - 8 | 964 | 95 | |
NM_001321858.1 | NP_001308787.1 | 6 - 7 | 715 | 95 | |
NM_003470.2 | NP_003461.2 | 6 - 7 | 920 | 95 | |
NR_135826.1 | 6 - 7 | 743 | 95 | ||
XM_017023652.1 | XP_016879141.1 | 6 - 7 | 1348 | 95 | |
XM_017023653.1 | XP_016879142.1 | 6 - 7 | 784 | 95 | |
GenBank mRNA | AK302771.1 | - | 6 - 7 | 721 | 95 |
AK302872.1 | - | 6 - 7 | 743 | 95 | |
AK302912.1 | - | 7 - 8 | 908 | 95 | |
AK316441.1 | - | 6 - 7 | 790 | 95 | |
AY376241.1 | - | 6 - 7 | 751 | 95 | |
Z72499.1 | - | 6 - 7 | 920 | 95 |