Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
Chromosome Location
Chr.17: 60677811 - 61392838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099432.1 NP_001092902.1 4 - 5 318 161
NM_001320470.1 NP_001307399.1 4 - 5 318 161
NM_017679.3 NP_060149.3 4 - 5 318 161
XM_005257472.1 XP_005257529.1 4 - 5 269 161
XM_011524939.1 XP_011523241.1 4 - 5 269 161
XM_011524940.1 XP_011523242.1 4 - 5 269 161
XM_011524941.1 XP_011523243.1 4 - 5 269 161
XM_011524942.2 XP_011523244.1 4 - 5 269 161
XM_011524943.2 XP_011523245.1 4 - 5 269 161
XM_011524944.1 XP_011523246.1 4 - 5 269 161
XM_011524945.2 XP_011523247.1 4 - 5 269 161
XM_011524946.2 XP_011523248.1 4 - 5 269 161
XM_011524947.1 XP_011523249.1 4 - 5 269 161
XM_011524948.2 XP_011523250.1 4 - 5 269 161
XM_011524949.2 XP_011523251.1 4 - 5 269 161
XM_011524950.1 XP_011523252.1 4 - 5 269 161
XM_011524951.2 XP_011523253.1 4 - 5 269 161
XM_017024783.1 XP_016880272.1 4 - 5 269 161
XM_017024784.1 XP_016880273.1 4 - 5 269 161
XM_017024785.1 XP_016880274.1 4 - 5 269 161
XM_017024786.1 XP_016880275.1 4 - 5 269 161
XM_017024787.1 XP_016880276.1 4 - 5 269 161
XM_017024788.1 XP_016880277.1 4 - 5 269 161
XM_017024789.1 XP_016880278.1 4 - 5 269 161
XM_017024790.1 XP_016880279.1 4 - 5 269 161
XM_017024791.1 XP_016880280.1 4 - 5 269 161
XM_017024792.1 XP_016880281.1 4 - 5 269 161
XM_017024793.1 XP_016880282.1 4 - 5 269 161
XM_017024794.1 XP_016880283.1 4 - 5 269 161
XM_017024795.1 XP_016880284.1 4 - 5 269 161
GenBank mRNA AF361219.1 - 5 - 6 257 161
AK025510.1 - 4 - 5 234 161
AK225757.1 - 4 - 5 277 161
AK314744.1 - 4 - 5 263 161
BC017390.1 - 4 - 5 274 161
BC117275.1 - 4 - 5 240 161
BC143386.1 - 4 - 5 240 161
BC143387.1 - 4 - 5 240 161
DA188234.1 - 4 - 5 319 161
DA491709.1 - 4 - 5 255 161
DA789232.1 - 4 - 5 283 161

View Full Product Details