Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BCAS3
Entrez Gene ID
54828
Gene Name
BCAS3, microtubule associated cell migration factor
Gene Aliases
GAOB1, MAAB
Chromosome Location
Chr.17: 60677811 - 61392838 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001099432.1 NP_001092902.1 6 - 7 509 129
NM_001320470.1 NP_001307399.1 6 - 7 509 129
NM_017679.3 NP_060149.3 6 - 7 509 129
XM_005257472.1 XP_005257529.1 6 - 7 460 129
XM_011524939.1 XP_011523241.1 6 - 7 460 129
XM_011524940.1 XP_011523242.1 6 - 7 460 129
XM_011524941.1 XP_011523243.1 6 - 7 460 129
XM_011524942.2 XP_011523244.1 6 - 7 460 129
XM_011524943.2 XP_011523245.1 6 - 7 460 129
XM_011524944.1 XP_011523246.1 6 - 7 460 129
XM_011524945.2 XP_011523247.1 6 - 7 460 129
XM_011524946.2 XP_011523248.1 6 - 7 460 129
XM_011524947.1 XP_011523249.1 6 - 7 460 129
XM_011524948.2 XP_011523250.1 6 - 7 460 129
XM_011524949.2 XP_011523251.1 6 - 7 460 129
XM_011524950.1 XP_011523252.1 6 - 7 460 129
XM_011524951.2 XP_011523253.1 6 - 7 460 129
XM_017024783.1 XP_016880272.1 6 - 7 460 129
XM_017024784.1 XP_016880273.1 6 - 7 460 129
XM_017024785.1 XP_016880274.1 6 - 7 460 129
XM_017024786.1 XP_016880275.1 6 - 7 460 129
XM_017024787.1 XP_016880276.1 6 - 7 460 129
XM_017024788.1 XP_016880277.1 6 - 7 460 129
XM_017024789.1 XP_016880278.1 6 - 7 460 129
XM_017024790.1 XP_016880279.1 6 - 7 460 129
XM_017024791.1 XP_016880280.1 6 - 7 460 129
XM_017024792.1 XP_016880281.1 6 - 7 460 129
XM_017024793.1 XP_016880282.1 6 - 7 460 129
XM_017024794.1 XP_016880283.1 6 - 7 460 129
XM_017024795.1 XP_016880284.1 6 - 7 460 129
GenBank mRNA AF361219.1 - 7 - 8 448 129
AK025510.1 - 6 - 7 425 129
AK225757.1 - 6 - 7 468 129
AK314744.1 - 6 - 7 454 129
BC017390.1 - 6 - 7 465 129
BC117275.1 - 6 - 7 431 129
BC143386.1 - 6 - 7 431 129
BC143387.1 - 6 - 7 431 129
DA491709.1 - 6 - 7 446 129
DA789232.1 - 6 - 7 474 129

View Full Product Details