The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015320.3 | NP_056135.2 | 9 - 10 | 1855 | 61 |
NM_032995.2 | NP_127462.1 | 8 - 9 | 1444 | 61 | |
XM_005263681.1 | XP_005263738.1 | 9 - 10 | 4932 | 61 | |
XM_005263687.3 | XP_005263744.1 | 6 - 7 | 1146 | 61 | |
XM_005263688.2 | XP_005263745.1 | 5 - 6 | 1076 | 61 | |
XM_005263689.4 | XP_005263746.1 | 6 - 7 | 991 | 61 | |
XM_011511274.2 | XP_011509576.1 | 9 - 10 | 4932 | 61 | |
XM_011511276.2 | XP_011509578.1 | 5 - 6 | 794 | 61 | |
XM_011511277.1 | XP_011509579.1 | 5 - 6 | 921 | 61 | |
GenBank mRNA | AB029035.1 | - | 8 - 9 | 1410 | 61 |
AB042199.1 | - | 6 - 7 | 1135 | 61 | |
AF249745.1 | - | 9 - 10 | 1856 | 61 | |
AL137289.1 | - | 4 - 5 | 756 | 61 |