The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145055.1 | NP_001138527.1 | 2 - 3 | 294 | 102 |
GenBank mRNA | AF110800.1 | - | 2 - 3 | 295 | 102 |