The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 10 - 11 | 1337 | 110 |
NM_003640.4 | NP_003631.2 | 10 - 11 | 1482 | 110 | |
XM_011519136.1 | XP_011517438.1 | 10 - 11 | 1274 | 110 | |
XM_011519137.1 | XP_011517439.1 | 10 - 11 | 1048 | 110 | |
XM_017015238.1 | XP_016870727.1 | 8 - 9 | 1053 | 110 | |
GenBank mRNA | AB210033.1 | - | 10 - 11 | 1244 | 110 |
AF044195.1 | - | 10 - 11 | 1261 | 110 | |
AF153419.2 | - | 10 - 11 | 1268 | 110 | |
AK001641.1 | - | 10 - 11 | 1266 | 110 | |
AK127237.1 | - | 2 - 3 | 897 | 110 | |
AK289962.1 | - | 10 - 11 | 1274 | 110 | |
BC033094.1 | - | 10 - 11 | 1044 | 110 | |
CR749385.1 | - | 3 - 4 | 309 | 110 |