The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 13 - 14 | 1830 | 99 |
NM_003640.4 | NP_003631.2 | 13 - 14 | 1975 | 99 | |
XM_011519136.1 | XP_011517438.1 | 13 - 14 | 1767 | 99 | |
XM_011519137.1 | XP_011517439.1 | 13 - 14 | 1541 | 99 | |
XM_017015238.1 | XP_016870727.1 | 11 - 12 | 1546 | 99 | |
GenBank mRNA | AB210033.1 | - | 13 - 14 | 1737 | 99 |
AF044195.1 | - | 13 - 14 | 1754 | 99 | |
AF153419.2 | - | 13 - 14 | 1761 | 99 | |
AK001641.1 | - | 13 - 14 | 1759 | 99 | |
AK127237.1 | - | 5 - 6 | 1390 | 99 | |
AK289962.1 | - | 13 - 14 | 1767 | 99 | |
AK304742.1 | - | 13 - 14 | 1472 | 99 | |
BC033094.1 | - | 13 - 14 | 1537 | 99 | |
CR749385.1 | - | 6 - 7 | 802 | 99 |