Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKBKAP
Entrez Gene ID
8518
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
Chromosome Location
Chr.9: 108867520 - 108934332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318360.1 NP_001305289.1 20 - 21 2576 86
NM_003640.4 NP_003631.2 20 - 21 2721 86
XM_011519136.1 XP_011517438.1 20 - 21 2513 86
XM_011519137.1 XP_011517439.1 20 - 21 2287 86
XM_017015238.1 XP_016870727.1 18 - 19 2292 86
GenBank mRNA AB210033.1 - 20 - 21 2483 86
AF044195.1 - 20 - 21 2500 86
AF153419.2 - 20 - 21 2507 86
AK001641.1 - 20 - 21 2505 86
AK022559.1 - 2 - 3 189 86
AK127237.1 - 12 - 13 2136 86
AK225798.1 - 7 - 8 805 86
AK289962.1 - 20 - 21 2513 86
AK304742.1 - 20 - 21 2218 86
BC033094.1 - 20 - 21 2283 86
CR749385.1 - 13 - 14 1548 86

View Full Product Details