The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318360.1 | NP_001305289.1 | 23 - 24 | 2887 | 125 |
NM_003640.4 | NP_003631.2 | 23 - 24 | 3032 | 125 | |
XM_011519136.1 | XP_011517438.1 | 23 - 24 | 2824 | 125 | |
XM_011519137.1 | XP_011517439.1 | 23 - 24 | 2598 | 125 | |
XM_017015238.1 | XP_016870727.1 | 21 - 22 | 2603 | 125 | |
GenBank mRNA | AB210033.1 | - | 23 - 24 | 2794 | 125 |
AF044195.1 | - | 23 - 24 | 2811 | 125 | |
AF153419.2 | - | 23 - 24 | 2818 | 125 | |
AK001641.1 | - | 23 - 24 | 2816 | 125 | |
AK024223.1 | - | 3 - 4 | 275 | 125 | |
AK127237.1 | - | 15 - 16 | 2447 | 125 | |
AK225798.1 | - | 10 - 11 | 1116 | 125 | |
AK289962.1 | - | 23 - 24 | 2824 | 125 | |
AK304742.1 | - | 23 - 24 | 2529 | 125 | |
BC033094.1 | - | 23 - 24 | 2594 | 125 | |
CR749385.1 | - | 16 - 17 | 1859 | 125 |