Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
IKBKAP
Entrez Gene ID
8518
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
Gene Aliases
DYS, ELP1, FD, IKAP, IKI3, TOT1
Chromosome Location
Chr.9: 108867520 - 108934332 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001318360.1 NP_001305289.1 4 - 5 762 113
NM_003640.4 NP_003631.2 4 - 5 907 113
XM_011519136.1 XP_011517438.1 4 - 5 699 113
XM_011519137.1 XP_011517439.1 4 - 5 473 113
XM_017015238.1 XP_016870727.1 4 - 5 666 113
GenBank mRNA AB210033.1 - 4 - 5 669 113
AF044195.1 - 4 - 5 686 113
AF153419.2 - 4 - 5 693 113
AK001641.1 - 4 - 5 691 113
AK289962.1 - 4 - 5 699 113
AK304742.1 - 4 - 5 404 113
BC033094.1 - 4 - 5 469 113

View Full Product Details