Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM26
Entrez Gene ID
64062
Gene Name
RNA binding motif protein 26
Gene Aliases
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location
Chr.13: 79311827 - 79406265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286631.1 NP_001273560.1 10 - 11 1991 70
NM_001286632.1 NP_001273561.1 10 - 11 1976 70
NM_022118.4 NP_071401.3 10 - 11 1976 70
XM_005266491.4 XP_005266548.1 10 - 11 2058 70
XM_005266497.1 XP_005266554.1 10 - 11 2058 70
XM_006719857.1 XP_006719920.1 10 - 11 2058 70
XM_011535179.1 XP_011533481.1 10 - 11 2058 70
XM_011535180.2 XP_011533482.1 10 - 11 2058 70
XM_011535181.1 XP_011533483.1 10 - 11 2058 70
XM_011535182.1 XP_011533484.1 10 - 11 2043 70
XM_011535183.2 XP_011533485.1 10 - 11 2058 70
XM_011535184.1 XP_011533486.1 10 - 11 2028 70
XM_011535185.2 XP_011533487.1 10 - 11 2028 70
XM_011535186.2 XP_011533488.1 10 - 11 2020 70
XM_011535187.2 XP_011533489.1 10 - 11 2028 70
XM_011535188.2 XP_011533490.1 10 - 11 2020 70
XM_011535189.1 XP_011533491.1 10 - 11 2058 70
XM_011535190.1 XP_011533492.1 10 - 11 2058 70
XM_011535191.2 XP_011533493.1 10 - 11 2058 70
XM_011535192.2 XP_011533494.1 10 - 11 2020 70
XM_011535193.2 XP_011533495.1 10 - 11 2013 70
XM_011535195.1 XP_011533497.1 10 - 11 1756 70
XM_017020688.1 XP_016876177.1 10 - 11 2058 70
XM_017020689.1 XP_016876178.1 10 - 11 2043 70
XM_017020690.1 XP_016876179.1 10 - 11 2050 70
XM_017020691.1 XP_016876180.1 10 - 11 2050 70
XM_017020692.1 XP_016876181.1 10 - 11 2020 70
XM_017020693.1 XP_016876182.1 10 - 11 2058 70
XM_017020694.1 XP_016876183.1 10 - 11 2043 70
XM_017020695.1 XP_016876184.1 10 - 11 2043 70
XM_017020696.1 XP_016876185.1 10 - 11 2050 70
XM_017020697.1 XP_016876186.1 10 - 11 2050 70
XM_017020698.1 XP_016876187.1 10 - 11 2013 70
XM_017020699.1 XP_016876188.1 10 - 11 2035 70
XM_017020700.1 XP_016876189.1 10 - 11 2020 70
XM_017020701.1 XP_016876190.1 10 - 11 2058 70
XM_017020702.1 XP_016876191.1 10 - 11 2058 70
XM_017020703.1 XP_016876192.1 10 - 11 2028 70
GenBank mRNA AK027339.1 - 5 - 6 759 70
AK027456.1 - 4 - 5 625 70
AK302871.1 - 10 - 11 1585 70
AL834343.1 - 5 - 6 889 70
AX775885.1 - 4 - 5 625 70
BC041655.1 - 10 - 11 1543 70
BC111697.1 - 10 - 11 1537 70
BC111739.1 - 10 - 11 1553 70
BX648672.1 - 10 - 11 1534 70

View Full Product Details