Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
RBM26
Entrez Gene ID
64062
Gene Name
RNA binding motif protein 26
Gene Aliases
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
Chromosome Location
Chr.13: 79311827 - 79406265 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001286631.1 NP_001273560.1 20 - 21 3275 149
NM_001286632.1 NP_001273561.1 19 - 20 3197 149
NM_022118.4 NP_071401.3 19 - 20 3188 149
XM_005266491.4 XP_005266548.1 20 - 21 3351 149
XM_005266497.1 XP_005266554.1 20 - 21 3351 149
XM_006719857.1 XP_006719920.1 20 - 21 3351 149
XM_011535179.1 XP_011533481.1 20 - 21 3351 149
XM_011535180.2 XP_011533482.1 20 - 21 3351 149
XM_011535181.1 XP_011533483.1 20 - 21 3342 149
XM_011535182.1 XP_011533484.1 20 - 21 3336 149
XM_011535183.2 XP_011533485.1 20 - 21 3342 149
XM_011535184.1 XP_011533486.1 20 - 21 3321 149
XM_011535185.2 XP_011533487.1 20 - 21 3321 149
XM_011535186.2 XP_011533488.1 20 - 21 3313 149
XM_011535187.2 XP_011533489.1 20 - 21 3312 149
XM_011535188.2 XP_011533490.1 20 - 21 3313 149
XM_011535189.1 XP_011533491.1 19 - 20 3279 149
XM_011535190.1 XP_011533492.1 19 - 20 3270 149
XM_011535191.2 XP_011533493.1 19 - 20 3270 149
XM_011535192.2 XP_011533494.1 19 - 20 3241 149
XM_011535193.2 XP_011533495.1 19 - 20 3225 149
XM_011535195.1 XP_011533497.1 20 - 21 3049 149
XM_017020688.1 XP_016876177.1 20 - 21 3351 149
XM_017020689.1 XP_016876178.1 20 - 21 3327 149
XM_017020690.1 XP_016876179.1 20 - 21 3343 149
XM_017020691.1 XP_016876180.1 20 - 21 3334 149
XM_017020692.1 XP_016876181.1 20 - 21 3304 149
XM_017020693.1 XP_016876182.1 19 - 20 3279 149
XM_017020694.1 XP_016876183.1 19 - 20 3264 149
XM_017020695.1 XP_016876184.1 19 - 20 3255 149
XM_017020696.1 XP_016876185.1 19 - 20 3271 149
XM_017020697.1 XP_016876186.1 20 - 21 3343 149
XM_017020698.1 XP_016876187.1 19 - 20 3234 149
XM_017020699.1 XP_016876188.1 19 - 20 3256 149
XM_017020700.1 XP_016876189.1 19 - 20 3232 149
GenBank mRNA AF273052.1 - 6 - 7 718 149
AK024610.1 - 4 - 5 547 149
AK027339.1 - 14 - 15 1971 149
AK027456.1 - 13 - 14 1846 149
AK302871.1 - 19 - 20 2806 149
AK302927.1 - 7 - 8 1085 149
AL834343.1 - 15 - 16 2182 149
AX775885.1 - 13 - 14 1846 149
BC000791.2 - 4 - 5 573 149
BC041655.1 - 19 - 20 2755 149
BC111697.1 - 19 - 20 2758 149
BC111739.1 - 20 - 21 2837 149

View Full Product Details