Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- RBM26
- Entrez Gene ID
-
64062
- Gene Name
- RNA binding motif protein 26
- Gene Aliases
-
ARRS2, C13orf10, PPP1R132, PRO1777, SE70-2, ZC3H17
- Chromosome Location
-
Chr.13: 79311827 - 79406265 on Build GRCh38
View Full Product Details