The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_040243.1 | 7 - 8 | 1477 | 149 | |
XM_006722340.3 | XP_006722403.1 | 7 - 8 | 1030 | 149 | |
XM_006722341.3 | XP_006722404.1 | 9 - 10 | 1652 | 149 | |
XM_006722342.3 | XP_006722405.1 | 8 - 9 | 1436 | 149 | |
XM_006722343.3 | XP_006722406.1 | 8 - 9 | 1557 | 149 | |
XM_006722345.3 | XP_006722408.1 | 6 - 7 | 1370 | 149 | |
XM_006722348.2 | XP_006722411.1 | 7 - 8 | 1030 | 149 | |
XM_011525731.2 | XP_011524033.1 | 10 - 11 | 2060 | 149 | |
XM_011525732.1 | XP_011524034.1 | 7 - 8 | 1030 | 149 | |
XM_017025920.1 | XP_016881409.1 | 9 - 10 | 1965 | 149 | |
XM_017025921.1 | XP_016881410.1 | 6 - 7 | 872 | 149 | |
XM_017025931.1 | XP_016881420.1 | 7 - 8 | 1030 | 149 | |
GenBank mRNA | AK309948.1 | - | 7 - 8 | 995 | 149 |
BC073994.1 | - | 7 - 8 | 989 | 149 |