Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIK3CG
Entrez Gene ID
5294
Gene Name
phosphatidylinositol-4,5-bisphosphate 3-kinase catalytic subunit gamma
Gene Aliases
PI3CG, PI3K, PI3Kgamma, PIK3, p110gamma, p120-PI3K
Chromosome Location
Chr.7: 106865278 - 106908978 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001282426.1 NP_001269355.1 8 - 9 2921 86
NM_001282427.1 NP_001269356.1 8 - 9 2845 86
NM_002649.3 NP_002640.2 8 - 9 3070 86
XM_005250443.3 XP_005250500.1 8 - 9 2938 86
XM_011516316.1 XP_011514618.1 8 - 9 2903 86
XM_017012328.1 XP_016867817.1 8 - 9 2903 86
GenBank mRNA AF327656.1 - 7 - 8 2760 86
AK292684.1 - 8 - 9 2845 86
AK313753.1 - 8 - 9 2828 86
BC035683.1 - 8 - 9 2813 86
BX648341.1 - 8 - 9 2903 86
X83368.1 - 8 - 9 3080 86

View Full Product Details