Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPLX2
Entrez Gene ID
10814
Gene Name
complexin 2
Gene Aliases
921-L, CPX-2, CPX2, Hfb1
Chromosome Location
Chr.5: 175796539 - 175884021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006650.3 NP_006641.1 2 - 3 258 97

View Full Product Details