The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291501.1 | NP_001278430.1 | 7 - 8 | 1304 | 144 |
NM_148674.4 | NP_683515.4 | 7 - 8 | 1304 | 144 | |
XM_011530144.2 | XP_011528446.1 | 7 - 8 | 1165 | 144 | |
XM_011530145.2 | XP_011528447.1 | 7 - 8 | 1356 | 144 | |
GenBank mRNA | AJ504806.2 | - | 7 - 8 | 1304 | 144 |
BC126208.1 | - | 7 - 8 | 1284 | 144 |