The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | XM_006722341.3 | XP_006722404.1 | 2 - 3 | 567 | 69 |
XM_006722343.3 | XP_006722406.1 | 2 - 3 | 567 | 69 | |
XM_011525731.2 | XP_011524033.1 | 2 - 3 | 703 | 69 | |
XM_017025920.1 | XP_016881409.1 | 2 - 3 | 703 | 69 | |
XM_017025922.1 | XP_016881411.1 | 2 - 3 | 567 | 69 | |
XM_017025923.1 | XP_016881412.1 | 2 - 3 | 703 | 69 | |
XM_017025925.1 | XP_016881414.1 | 2 - 3 | 567 | 69 | |
XM_017025926.1 | XP_016881415.1 | 2 - 3 | 703 | 69 | |
XM_017025928.1 | XP_016881417.1 | 2 - 3 | 567 | 69 |