The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018420.2 | NP_060890.2 | 5 - 6 | 865 | 67 |
XM_005271004.3 | XP_005271061.1 | 5 - 6 | 826 | 67 | |
GenBank mRNA | AK125072.1 | - | 3 - 4 | 433 | 67 |
AY145501.1 | - | 5 - 6 | 735 | 67 | |
BC026358.1 | - | 5 - 6 | 864 | 67 | |
KU178698.1 | - | 5 - 6 | 699 | 67 | |
KU178700.1 | - | 5 - 6 | 699 | 67 | |
KU178703.1 | - | 4 - 5 | 486 | 67 |