The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001105203.1 | NP_001098673.1 | 5 - 6 | 1939 | 121 |
NM_001105204.1 | NP_001098674.1 | 5 - 6 | 1939 | 121 | |
NM_001105205.1 | NP_001098675.1 | 4 - 5 | 961 | 121 | |
NM_001278227.1 | NP_001265156.1 | 4 - 5 | 905 | 121 | |
NM_001278228.1 | NP_001265157.1 | 3 - 4 | 1141 | 121 | |
NM_001278229.1 | NP_001265158.1 | 4 - 5 | 1004 | 121 | |
NM_001278230.1 | NP_001265159.1 | 3 - 4 | 803 | 121 | |
NM_014328.4 | NP_055143.2 | 4 - 5 | 1004 | 121 | |
NR_103478.1 | 3 - 4 | 669 | 121 | ||
XM_006711254.1 | XP_006711317.1 | 6 - 7 | 2350 | 121 | |
XM_006711256.1 | XP_006711319.1 | 6 - 7 | 2350 | 121 | |
XM_006711257.1 | XP_006711320.1 | 4 - 5 | 618 | 121 | |
XM_017000891.1 | XP_016856380.1 | 5 - 6 | 2520 | 121 | |
XM_017000892.1 | XP_016856381.1 | 4 - 5 | 618 | 121 | |
GenBank mRNA | AB026894.2 | - | 4 - 5 | 428 | 121 |
AK055451.1 | - | 4 - 5 | 1199 | 121 | |
AK074485.1 | - | 3 - 4 | 557 | 121 | |
AK074982.1 | - | 4 - 5 | 423 | 121 | |
AK093777.1 | - | 3 - 4 | 594 | 121 | |
AK125378.1 | - | 5 - 6 | 1891 | 121 | |
AK298730.1 | - | 4 - 5 | 415 | 121 | |
AK307736.1 | - | 3 - 4 | 591 | 121 | |
AK314559.1 | - | 4 - 5 | 673 | 121 | |
AL080083.1 | - | 3 - 4 | 281 | 121 | |
BC001045.2 | - | 4 - 5 | 445 | 121 | |
BC004293.1 | - | 3 - 4 | 525 | 121 | |
BC028055.1 | - | 3 - 4 | 553 | 121 | |
BC052277.1 | - | 4 - 5 | 961 | 121 | |
BX402179.2 | - | 4 - 5 | 495 | 121 | |
BX640612.1 | - | 3 - 4 | 621 | 121 | |
CR749671.1 | - | 3 - 4 | 1510 | 121 |