The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172623.1 | NP_001166094.1 | 18 - 19 | 3050 | 120 |
NM_001172624.1 | NP_001166095.1 | 18 - 19 | 3050 | 120 | |
NM_002499.3 | NP_002490.2 | 18 - 19 | 3050 | 120 | |
XM_005254408.1 | XP_005254465.1 | 19 - 20 | 3360 | 120 | |
XM_011521628.2 | XP_011519930.1 | 18 - 19 | 3115 | 120 | |
XM_011521629.2 | XP_011519931.1 | 18 - 19 | 3109 | 120 | |
XM_011521630.2 | XP_011519932.1 | 18 - 19 | 3061 | 120 | |
XM_011521632.2 | XP_011519934.1 | 18 - 19 | 3109 | 120 | |
XM_011521633.2 | XP_011519935.1 | 18 - 19 | 3109 | 120 | |
XM_017022231.1 | XP_016877720.1 | 18 - 19 | 3001 | 120 | |
XM_017022232.1 | XP_016877721.1 | 19 - 20 | 3312 | 120 | |
XM_017022233.1 | XP_016877722.1 | 18 - 19 | 3109 | 120 | |
XM_017022234.1 | XP_016877723.1 | 19 - 20 | 3104 | 120 | |
XM_017022235.1 | XP_016877724.1 | 18 - 19 | 3061 | 120 | |
XM_017022236.1 | XP_016877725.1 | 18 - 19 | 3049 | 120 | |
XM_017022237.1 | XP_016877726.1 | 18 - 19 | 3109 | 120 | |
XM_017022238.1 | XP_016877727.1 | 18 - 19 | 3001 | 120 | |
GenBank mRNA | AB209412.1 | - | 14 - 15 | 1898 | 120 |
BC117161.1 | - | 18 - 19 | 2924 | 120 | |
BC143270.1 | - | 18 - 19 | 2925 | 120 | |
BC143271.1 | - | 18 - 19 | 2924 | 120 | |
U61262.1 | - | 18 - 19 | 2994 | 120 | |
U72391.1 | - | 18 - 19 | 2984 | 120 |