Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PLEKHO2
Entrez Gene ID
80301
Gene Name
pleckstrin homology domain containing O2
Gene Aliases
PLEKHQ1, PP1628, pp9099
Chromosome Location
Chr.15: 64841883 - 64868002 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025201.4 NP_079477.2 1 - 2 145 128
GenBank mRNA AF494534.1 - 1 - 2 94 128
AK096203.1 - 1 - 2 113 128

View Full Product Details