The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025201.4 | NP_079477.2 | 2 - 3 | 298 | 95 |
GenBank mRNA | AF494534.1 | - | 2 - 3 | 247 | 95 |
AK096203.1 | - | 2 - 3 | 266 | 95 |