The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001286490.1 | NP_001273419.1 | 5 - 6 | 519 | 73 |
NM_001322949.1 | NP_001309878.1 | 4 - 5 | 569 | 73 | |
NM_001322950.1 | NP_001309879.1 | 4 - 5 | 569 | 73 | |
NM_001322951.1 | NP_001309880.1 | 3 - 4 | 464 | 73 | |
NM_032892.4 | NP_116281.2 | 4 - 5 | 569 | 73 | |
XM_005254730.3 | XP_005254787.1 | 4 - 5 | 474 | 73 | |
GenBank mRNA | AK090572.1 | - | 4 - 5 | 501 | 73 |
AK123018.1 | - | 5 - 6 | 519 | 73 | |
BC053647.1 | - | 4 - 5 | 506 | 73 |