Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PSRC1
Entrez Gene ID
84722
Gene Name
proline and serine rich coiled-coil 1
Gene Aliases
DDA3, FP3214
Chromosome Location
Chr.1: 109279554 - 109283182 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001032291.2 NP_001027462.1 5 - 6 781 103
NM_032636.7 NP_116025.1 5 - 6 776 103
XM_005271283.2 XP_005271340.1 5 - 6 775 103
XM_017002568.1 XP_016858057.1 4 - 5 1083 103
XM_017002569.1 XP_016858058.1 5 - 6 788 103
XM_017002570.1 XP_016858059.1 5 - 6 792 103
GenBank mRNA AF223000.1 - 5 - 6 755 103
AF447874.1 - 2 - 3 919 103
AK289593.1 - 5 - 6 718 103

View Full Product Details