The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006187.3 | NP_006178.2 | 11 - 12 | 2506 | 68 |
XM_005253889.3 | XP_005253946.1 | 11 - 12 | 2427 | 68 | |
GenBank mRNA | AB044545.1 | - | 11 - 12 | 2587 | 68 |
AF063613.1 | - | 11 - 12 | 2442 | 68 | |
AK292949.1 | - | 11 - 12 | 2476 | 68 | |
BC113746.1 | - | 11 - 12 | 2481 | 68 |